missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford L-Cystine
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
CAS Number: 56-89-3
Molecular Formula: C6H12N2O4S2
Molecular Weight: 240.3
Chemical Purity: >98.5%
Soluble in:?1M HCl
Store at: Room TemperatureSpecifications
Specifications
| Chemical Name or Material | L-Cystine |
| CAS | 56-89-3 |
| Molecular Formula | C6H12N2O4S2 |
| Quantity | 5 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction