missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteine, MP Biomedicals™

Product Code. 11433400 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433400 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11433400 Supplier MP Biomedicals Supplier No. 0210144405
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 52-90-4
Color White
Molecular Formula C3H7NO2S
MDL Number MFCD00064306
Synonym l-cysteine, cysteine, cystein, half-cystine, r-cysteine, thioserine, r-2-amino-3-mercaptopropanoic acid, l-+-cysteine, l-cystein, 2r-2-amino-3-sulfanylpropanoic acid
InChI Key XUJNEKJLAYXESH-REOHCLBHSA-N
Density 1.334g/cm3(L)
SMILES C(C(C(=O)O)N)S
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid
Molecular Weight (g/mol) 121.154
Quantity 5 g
PubChem CID 5862
ChEBI CHEBI:17561
Formula Weight 121.16
Physical Form Solid
Chemical Name or Material L-Cysteine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.