missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteine methyl ester hydrochloride, MedChemExpress

Product Code. 16402309
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16402309 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16402309 Supplier Medchem Express Supplier No. HYB1038
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-Cysteine methyl ester hydrochloride is an antitussive, and an expectorant agent, used to relieve breathing difficulties caused by mucus.

L-Cysteine methyl ester hydrochloride is an antitussive, and an expectorant agent, used to relieve breathing difficulties caused by mucus.

Specifications

Chemical Name or Material L-Cysteine methyl ester hydrochloride
CAS 18598-63-5
Color White
Purity Grade Notes Research
Molecular Formula C4H10ClNO2S
Quantity 1 g
Solubility Information DMSO : 100 mg/mL (582.58 mM; Need ultrasonic)
SMILES N[C@@H](CS)C(OC)=O.[H]Cl
Molecular Weight (g/mol) 171.65
Formula Weight 171.65
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.