missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Cysteine Hydrochloride Monohydrate, 98.5 to 101.5% (Dry Basis)
Shop All Thermo Scientific ProductsSpecifications
Specifications
| Melting Point | ∼175°C (decomposition) |
| Packaging | Plastic Bottle |
| Linear Formula | HSCH2CH(NH2)COOH.HCl.H2O |
| Beilstein | 5158059 |
| Quantity | 5 kg |
| Formula Weight | 175.63 |
| Percent Purity | 98.5 to 101.5% |
| Physical Form | Crystalline Powder |
| Assay | Dry basis |
| Chemical Name or Material | L-Cysteine Hydrochloride Monohydrate |
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?