missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Cysteine Ethyl Ester Hydrochloride, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | L-Cysteine Ethyl Ester Hydrochloride |
| CAS | 868-59-7 |
| Molecular Formula | C5H11NO2S.HCl |
| Quantity | 25 g |
| Synonym | Cystanin, Ethyl L-cysteinate hydrochloride, Ethyl cysteinate hydrochloride, NSC 117387, NSC 519837 |
| InChI Formula | InChI=1S/C5H11NO2S.ClH/c1-2-8-5(7)4(6)3-9;/h4,9H,2-3,6H2,1H3;1H/t4-;/m0./s1 |
| SMILES | Cl.CCOC(=O)[C@@H](N)CS |
| IUPAC Name | ethyl (2R)-2-amino-3-sulfanylpropanoate;hydrochloride |
| Molecular Weight (g/mol) | 185.67 |
| Formula Weight | 185.0277 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction