missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteine ethyl ester hydrochloride, 98+%

Product Code. p-7044119
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11301677

Brand: Thermo Scientific Alfa Aesar L04692.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 868-59-7 | C5H12ClNO2S | 185.666 g/mol

L-Cysteine ethyl ester hydrochloride is widely used in food additive, cosmetic, pharmaceutical area.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Cysteine ethyl ester hydrochloride is widely used in food additive, cosmetic, pharmaceutical area.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from moisture and oxidizing agents. Incompatible with oxidizing agents and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868-59-7
Molecular Formula C5H12ClNO2S
Molecular Weight (g/mol) 185.666
MDL Number MFCD00012631
InChI Key JFKJWWJOCJHMGV-WCCKRBBISA-N
Synonym l-cysteine ethyl ester hydrochloride, h-cys-oet.hcl, r-ethyl 2-amino-3-mercaptopropanoate hydrochloride, l-cystein ethyl ester hydrochloride, ethyl l-cysteinate hydrochloride, cysteine ethyl ester hydrochloride, l-cysteine ethyl ester hcl, unii-am467tb2i1, ethyl cysteinate hydrochloride, cysteine, ethyl ester, hydrochloride
PubChem CID 2723617
IUPAC Name ethyl (2R)-2-amino-3-sulfanylpropanoate;hydrochloride
SMILES CCOC(=O)C(CS)N.Cl

Specifications

Melting Point 123°C to 125°C
Beilstein 3562600
Solubility Information Soluble in water.
Quantity 25 g
Formula Weight 185.67
Optical Rotation −12° (c=1 in 1N HCl)
Sensitivity Hygroscopic
Percent Purity ≥98%
Chemical Name or Material L-Cysteine ethyl ester hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.