missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteic Acid Monohydrate, TRC

Product Code. 30134235
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134235 10 g 10g
30133347 50 g 50g
30006010 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134235 Supplier Toronto Research Chemicals Supplier No. TRCC99450010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23537-25-9
Molecular Formula C3 H7 N O5 S . H2 O
Molecular Weight (g/mol) 187.17
Synonym 3-Sulfo-L-alanine Monohydrate, (R)-2-Amino-3-sulfopropanoic Acid Monohydrate
IUPAC Name (2R)-2-amino-3-sulfopropanoic acid;hydrate
SMILES O.N[C@@H](CS(=O)(=O)O)C(=O)O

Specifications

InChI Formula InChI=1S/C3H7NO5S.H2O/c4-2(3(5)6)1-10(7,8)9;/h2H,1,4H2,(H,5,6)(H,7,8,9);1H2/t2-;/m0./s1
Quantity 10 g
Formula Weight 187.0151
Shipping Condition Room Temperature
Chemical Name or Material L-Cysteic Acid Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.