missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteic acid, 99%

Product Code. 11905741 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11905741

Brand: Thermo Scientific Acros 226120500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

CAS: 498-40-8 | C3H7NO5S | 169.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-40-8
Molecular Formula C3H7NO5S
Molecular Weight (g/mol) 169.15
MDL Number MFCD00007524
InChI Key XVOYSCVBGLVSOL-UHFFFAOYNA-N
Synonym l-cysteic acid, cysteinesulfonic acid, l-alanine, 3-sulfo, 3-sulfo-l-alanine, cysteic acid, l, r-2-amino-3-sulfopropanoic acid, cysteic acid van, l-cysteate, cysteinic acid, cysteric acid
PubChem CID 72886
ChEBI CHEBI:17285
SMILES NC(CS(O)(=O)=O)C(O)=O

Specifications

Melting Point 267.0°C to 269.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula HO3SCH2CH(NH2)CO2H
Quantity 50 g
Beilstein 04,533
Merck Index 15,2777
Specific Rotation 8.3
Solubility Information (5% in water) Clear colorless
IUPAC Name (2R)-2-amino-3-sulfopropanoic acid
Formula Weight 169.16
Percent Purity 99%
Physical Form Crystalline Powder or Powder
Chemical Name or Material L-Cysteic acid
Show More Show Less
compliance-icons
Product Identifier
  • L-Cysteic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.