missing translation for 'onlineSavingsMsg'
Learn More

L-Citrulline, MedChemExpress

Product Code. 16402529
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16402529 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16402529 Supplier Medchem Express Supplier No. HYN0391

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress L-Citrulline is an amino acid derived from ornithine in the catabolism of proline or glutamine and glutamate, or from l-arginine via arginine-citrulline pathway.

L-Citrulline is an amino acid derived from ornithine in the catabolism of proline or glutamine and glutamate, or from l-arginine via arginine-citrulline pathway.

Specifications

Chemical Name or Material L-Citrulline
CAS 372-75-8
Color White
Purity Grade Notes Research
Molecular Formula C6H13N3O3
Solubility Information H2O : ≥ 50 mg/mL (285.40 mM)
SMILES N[C@@H](CCCNC(N)=O)C(O)=O
Molecular Weight (g/mol) 175.19
Quantity 1 g
Formula Weight 175.19
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.