missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Carnosine, 98%

Product Code. 10239703 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10239703 100 g Glass bottle 100g
10311753 1 g Glass bottle 1g
10348440 5 g Glass bottle 5g
10697684 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10239703 Supplier Thermo Scientific Chemicals Supplier No. 208171000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 305-84-0 | C9H14N4O3 | 226.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 305-84-0
Molecular Formula C9H14N4O3
Molecular Weight (g/mol) 226.23
MDL Number MFCD00005207
InChI Key CQOVPNPJLQNMDC-ZETCQYMHSA-N
Synonym l-carnosine, carnosine, beta-alanyl-l-histidine, karnozin, karnozzn, ignotine, l-ignotine, n-2-m, unii-8ho6pvn24w, nalpha-beta-alanyl-l-histidine
PubChem CID 439224
ChEBI CHEBI:15727
IUPAC Name (2S)-2-(3-aminopropanoylamino)-3-(1H-imidazol-5-yl)propanoic acid
SMILES C1=C(NC=N1)CC(C(=O)O)NC(=O)CCN

Specifications

Melting Point 253.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Beilstein 25, 516
Merck Index 15, 1851
Specific Rotation Condition +20.90° (20°C c=1.5,H2O)
Specific Rotation + 20.90
Formula Weight 226.23
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material L-Carnosine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.