missing translation for 'onlineSavingsMsg'
Learn More

L-Carnitine, TRC

Product Code. 30132984
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132984 5 g 5g
17913545 1 g 1g
17933755 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132984 Supplier Toronto Research Chemicals Supplier No. TRCC1841105G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-15-1
Molecular Formula C7 H15 N O3
Molecular Weight (g/mol) 161.2
Synonym 1-Propanaminium, 3-carboxy-2-hydroxy-N,N,N-trimethyl-, hydroxide, inner salt, (R)-, Ammonium, (3-carboxy-2-hydroxypropyl)trimethyl-, hydroxide, inner salt, L- (8CI), (-)-Carnitine, (-)-L-Carnitine, (R)-Carnitine, 1-Propanaminium, 3-carboxy-2-hydroxy-N,N,N-trimethyl-, inner salt, (R)-, Aminocarnifarm, Carniking, Carniking 50, Carnil, Carnilean, Carnipass, Carnipass 20, Carnitene, Carnitine, Carnitine, (-)-, Carnitor, Carnivit, Coaxel, L-(-)-Carnitine, L-Carnipure, L-Carnitine, Levocarnitine, Natrulon RC, ST 198, Vitamin BT, l-Carnitine
IUPAC Name (3R)-3-hydroxy-4-(trimethylazaniumyl)butanoate
SMILES C[N+](C)(C)C[C@H](O)CC(=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H15NO3/c1-8(2,3)5-6(9)4-7(10)11/h6,9H,4-5H2,1-3H3/t6-/m1/s1
Formula Weight 161.1052
Percent Purity >95%
Chemical Name or Material L-Carnitine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.