missing translation for 'onlineSavingsMsg'
Learn More

L-Carnitine Orotate, TRC

Product Code. 30358995
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358995 500 mg -
30356593 100 mg -
30353569 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30358995 Supplier Toronto Research Chemicals Supplier No. TRCC184170500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 160468-17-7
Molecular Formula C12H17N3O6
Molecular Weight (g/mol) 299.28
Synonym (2R)-3-Carboxy-2-hydroxy-N,N,N-trimethyl-1-propanaminium-1,2,3,6-tetrahydro-2,6-dioxo-4-pyrimidinecarboxylic Acid Salt, (2R)-3-Carboxy-2-hydroxy-N,N,N-trimethyl-1-propanaminium-1,2,3,6-tetrahydro-2,6-dioxo-4-pyrimidinecarboxylic Acid Ion, (R)-3-Carboxy-2-hydroxy-N,N,N-trimethyl-1-propanaminium-1,2,3,6-tetrahydro-2,6-dioxo-4-pyrimidinecarboxylic Acid Ion, (R)- 3-Carboxy-2-hydroxy-N,N,N-trimethyl-1-propanaminium-1,2,3,6-tetrahydro-2,6-dioxo-4-pyrimidinecarboxylic Acid
IUPAC Name [(2R)-3-carboxy-2-hydroxypropyl]-trimethylazanium;2,4-dioxo-1H-pyrimidine-6-carboxylate
SMILES C[N+](C)(C)C[C@H](O)CC(=O)O.[O-]C(=O)C1=CC(=O)NC(=O)N1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C7H15NO3.C5H4N2O4/c1-8(2,3)5-6(9)4-7(10)11;8-3-1-2(4(9)10)6-5(11)7-3/h6,9H,4-5H2,1-3H3;1H,(H,9,10)(H2,6,7,8,11)/t6-;/m1./s1
Formula Weight 317.1223
Chemical Name or Material L-Carnitine Orotate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.