missing translation for 'onlineSavingsMsg'
Learn More

L-Carnitine-L-tartrate, TRC

Product Code. 30137410
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137410 500 mg 500mg
30012882 100 mg 100mg
30005026 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137410 Supplier Toronto Research Chemicals Supplier No. TRCC184173500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36687-82-8
Molecular Formula 2 C7 H16 N O3 . C4 H4 O6
Molecular Weight (g/mol) 472.48
Synonym 1-Propanaminium, 3-carboxy-2-hydroxy-N,N,N-trimethyl-, (2R)-, (2R,3R)-2,3-dihydroxybutanedioate (2:1), 1-Propanaminium, 3-carboxy-2-hydroxy-N,N,N-trimethyl-, (2R)-, salt with (2R,3R)-2,3-dihydroxybutanedioic acid (2:1) (9CI), 1-Propanaminium, 3-carboxy-2-hydroxy-N,N,N-trimethyl-, (R)-, salt with [R-(R*,R*)]-2,3-dihydroxybutanedioic acid (2:1), Butanedioic acid, 2,3-dihydroxy- (2R,3R)-, ion(2-), bis[(2R)-3-carboxy-2-hydroxy-N,N,N-trimethyl-1-propanaminium] (9CI), Butanedioic acid, 2,3-dihydroxy- [R-(R*,R*)]-, ion(2-), bis[(R)-3-carboxy-2-hydroxy-N,N,N-trimethyl-1-propanaminium], Carnipure tartrate, Carnitine tartrate, L-Carnitine L-tartrate, L-Carnitine tartrate, Levocarnitine tartrate, Levocarnitine L-Tartrate
IUPAC Name [(2R)-3-carboxy-2-hydroxypropyl]-trimethylazanium;(2R,3R)-2,3-dihydroxybutanedioate
SMILES C[N+](C)(C)C[C@H](O)CC(=O)O.C[N+](C)(C)C[C@H](O)CC(=O)O.O[C@H]([C@@H](O)C(=O)[O-])C(=O)[O-]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/2C7H15NO3.C4H6O6/c2*1-8(2,3)5-6(9)4-7(10)11;5-1(3(7)8)2(6)4(9)10/h2*6,9H,4-5H2,1-3H3;1-2,5-6H,(H,7,8)(H,9,10)/t2*6-;1-,2-/m111/s1
Formula Weight 472.2268
Percent Purity >95%
Chemical Name or Material L-Carnitinetartrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.