missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Carnitine hydrochloride, 98%

Product Code. 15215988 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15215988 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15215988 Supplier Thermo Scientific Acros Supplier No. 230280250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6645-46-1 | C7H16ClNO3 | 197.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6645-46-1
Melting Point 142.0°C
Loss on Drying 5% max.
Infrared Spectrum Authentic
Molecular Formula C7H16ClNO3
Linear Formula HO2CCH2CH(OH)CH2N(CH3)3Cl
MDL Number MFCD00066100
Quantity 25 g
Merck Index 15,1849
Specific Rotation Condition −22.00 (20.00°C c=1, H2O)
Specific Rotation -22
Synonym r-3-carboxy-2-hydroxy-n,n,n-trimethylpropan-1-aminium chloride, l-carnitine hcl, unii-j3y5e6iks3, l-carnitine hydrochloride, levocarnitine chloride, j3y5e6iks3, r-carnitine hydrochloride, carnitine l-form hydrochloride, +--carnitine chloride, l-3-carboxy-2-hydroxypropyl trimethylammonium chloride
Solubility Information Solubility in water: soluble. Other solubilities: soluble in hot ethanol, slightly soluble in cold ethanol, practically insoluble in acetone and ether
InChI Key JXXCENBLGFBQJM-FYZOBXCZSA-N
SMILES [Cl-].C[N+](C)(C)C[C@H](O)CC(O)=O
IUPAC Name [(2R)-3-carboxy-2-hydroxypropyl]-trimethylazanium;chloride
Molecular Weight (g/mol) 197.66
PubChem CID 656657
Formula Weight 197.66
Percent Purity 98%
Chemical Name or Material L(-)-Carnitine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • L(-)-Carnitine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.