missing translation for 'onlineSavingsMsg'
Learn More

L-Carnitine, 98+%

Product Code. 11478247
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11478247

Brand: Thermo Scientific Alfa Aesar A17618.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 541-15-1 | C7H15NO3 | 161.201 g/mol

L-Carnitine is widely used as a nutritional supplement in food and beverage industries. It is used as a food additive. It acts as a cyanide antidote. It is also used in reversing and preventing hyperthyroid symptoms. It is a vitamin-like nutrient, which plays an important role in human metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Carnitine is widely used as a nutritional supplement in food and beverage industries. It is used as a food additive. It acts as a cyanide antidote. It is also used in reversing and preventing hyperthyroid symptoms. It is a vitamin-like nutrient, which plays an important role in human metabolism.

Solubility
Soluble in water, pyridine, dimethyl sulfoxide, methanol and ethanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-15-1
Molecular Formula C7H15NO3
Molecular Weight (g/mol) 161.201
MDL Number MFCD00038747
InChI Key PHIQHXFUZVPYII-ZCFIWIBFSA-N
Synonym l-carnitine, levocarnitine, vitamin bt, r-carnitine, carnitor, --carnitine, carnitine, --l-carnitine, karnitin, l---carnitine
PubChem CID 10917
ChEBI CHEBI:16347
IUPAC Name (3R)-3-hydroxy-4-(trimethylazaniumyl)butanoate
SMILES C[N+](C)(C)CC(CC(=O)[O-])O

Specifications

Melting Point ∼190°C (decomposition)
Quantity 250 g
Beilstein 4292315
Sensitivity Hygroscopic
Merck Index 14,1849
Solubility Information Soluble in water,pyridine,dimethyl sulfoxide,methanol and ethanol.
Formula Weight 161.2
Percent Purity ≥99%
Chemical Name or Material L-Carnitine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.