missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Camphorsulfonic acid, 98%

Product Code. 10477192 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10477192 5 g Glass bottle 5g
10207600 25 g Glass bottle 25g
10561544 100 g Glass bottle 100g
13468488 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10477192 Supplier Thermo Scientific Acros Supplier No. 263640050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35963-20-3 | C10H16O4S | 232.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35963-20-3
Molecular Formula C10H16O4S
Molecular Weight (g/mol) 232.29
MDL Number MFCD00150753,MFCD00064158
InChI Key MIOPJNTWMNEORI-MHPPCMCBSA-N
Synonym l--camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, 1r---camphor-10-sulfonic acid, unii-y6075i4fxe, camphorsulfonic acid,-, l---camphor-10-sulfonic acid, s-camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 hept-1-yl methanesulfonic acid, --camphor-10-sulfonic acid
PubChem CID 5771688
ChEBI CHEBI:55401
SMILES CC1(C)C2CC[C@]1(CS(O)(=O)=O)C(=O)C2

Specifications

Melting Point 198.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 11,316
Fieser 16,58
Specific Rotation − 22.00
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in glacial acetic acid and ethyl,acetate,practically insoluble in ether,soluble in chloroform and alcohol
IUPAC Name [(1S,4R)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid
Formula Weight 232.3
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material L(-)-Camphorsulfonic acid
Show More Show Less
compliance-icons
Product Identifier
  • L(-)-Camphorsulfonic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.