missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ L-Buthionine sulfoximine

Description
L-Buthionine sulfoximine is an irreversible γ-glutamylcysteine synthetase (γ-GCS) inhibitor; depletes glutathione levels by inhibiting synthesis. Induces ferroptosis in vitro.
Specifications
Specifications
| Chemical Name or Material | (2S)-2-Amino-4-(S-butylsulfonimidoyl)butanoic acid |
| CAS | 83730-53-4 |
| Quantity | 100 mg |
| Solubility | Soluble to 100 mM in water and to 100 mM in DMSO |
| Molecular Formula | C8H18N2O3S |
| Formula Weight | 222.3 |
| Product Type | Inhibitor |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?