missing translation for 'onlineSavingsMsg'
Learn More

L-Buthionine-(S,R)-sulfoximine, TRC

Product Code. 30135035
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135035 1 g 1g
30005992 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135035 Supplier Toronto Research Chemicals Supplier No. TRCB6902701G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83730-53-4
Molecular Formula C8 H18 N2 O3 S
Molecular Weight (g/mol) 222.31
IUPAC Name (2S)-2-amino-4-(butylsulfonimidoyl)butanoic acid
SMILES CCCCS(=N)(=O)CC[C@H](N)C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H18N2O3S/c1-2-3-5-14(10,13)6-4-7(9)8(11)12/h7,10H,2-6,9H2,1H3,(H,11,12)/t7-,14?/m0/s1
Formula Weight 222.1038
Percent Purity >95%
Chemical Name or Material L-Buthionine-(S,R)-sulfoximine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.