missing translation for 'onlineSavingsMsg'
Learn More

L-Buthionine-(S,R)-sulfoximine, MedChemExpress

Product Code. 16490437
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490437 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16490437 Supplier Medchem Express Supplier No. HY106376A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress L-Buthionine-(S,R)-sulfoximine is a cell-permeable, potent, fast acting and irreversible inhibitor of g-glutamylcysteine synthetase and depletes cellular glutathione levels. The IC50 value of L-Buthionine-(S,R)-sulfoximine on melanoma, breast and ovarian tumor specimens are 1.9 μM, 8.6 μM, and 29 μM, respectively.

L-Buthionine-(S,R)-sulfoximine is a cell-permeable, potent, fast acting and irreversible inhibitor of g-glutamylcysteine synthetase and depletes cellular glutathione levels. The IC50 value of L-Buthionine-(S,R)-sulfoximine on melanoma, breast and ovarian tumor specimens are 1.9 μM, 8.6 μM, and 29 μM, respectively.

Specifications

Chemical Name or Material L-Buthionine-(S,R)-sulfoximine
Physical Form Solid
CAS 83730-53-4
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C8H18N2O3S
For Use With (Application) Cancer-programmed cell death
Synonym L-Buthionine sulfoximine L-BSO
Solubility Information H2O : 18.33 mg/mL (82.45 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (ultrasonic) (insoluble or slightly soluble)
SMILES N[C@H](C(O)=O)CCS(CCCC)(=O)=N
Molecular Weight (g/mol) 222.31
Formula Weight 222.31
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.