missing translation for 'onlineSavingsMsg'
Learn More

L-Buthionine-(S,R)-sulfoximine, 99%

Product Code. 10146873 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10146873 5 g Glass bottle 5g
10512831 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10146873 Supplier Thermo Scientific Acros Supplier No. 235520050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 83730-53-4 | C8H18N2O3S | 222.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83730-53-4
Molecular Formula C8H18N2O3S
Molecular Weight (g/mol) 222.31
MDL Number MFCD00067000
InChI Key KJQFBVYMGADDTQ-CVSPRKDYSA-N
Synonym l-buthionine-sulfoximine, l-buthionine sulfoximine, 2s-2-amino-4-butylsulfonimidoyl butanoic acid, l-buthionine-s,r-sulfoximine, l-buthionine s,r-sulfoximine, l-bso, butanoic acid, 2-amino-4-s-butylsulfonimidoyl-, 2s, l-buthionine sr-sulfoximine, buthionine-s,r-sulfoximine, l, l-buthionine-s,r-sulphoximine
PubChem CID 119565
IUPAC Name (2S)-2-amino-4-(butylsulfonimidoyl)butanoic acid
SMILES CCCCS(=N)(=O)CCC(C(=O)O)N

Specifications

Melting Point 234.0°C to 235.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HOOCCH(NH2)CH2CH2S(O)(=NH)(CH2)3CH3
Quantity 5 g
Specific Rotation Condition +33° (20°C c=1,1N HCl)
Specific Rotation + 33.00
Solubility Information Solubility in water: soluble
Formula Weight 222.31
Percent Purity 99%
Physical Form Fine Powder
Chemical Name or Material L-Buthionine-(S, R)-sulfoximine
Show More Show Less
compliance-icons
Product Identifier
  • L-Buthionine-(S,R)-sulfoximine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.