missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ L-Broth Agar, Large Capsules

Product Code. 11393209 Shop All MP Biomedicals Products
Click to view available options
:
Quantity: 1kg

Product Code. 11393209

Brand: MP Biomedicals™ 113001236

Please to purchase this item. Need a web account? Register with us today!

Culture media used for the growth of bacteria.

  • Typically contain 10g of tryptone and 5g of yeast extract per liter, and vary in salt
TRUSTED_SUSTAINABILITY

Specifications

Broth Type L-broth
Media Type Bacterial media
Product Type Agar
For Use With (Application) Bacteria cell culture, gene expression; microbiological analysis; transfection and protein expression
Quantity 1 kg

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.