missing translation for 'onlineSavingsMsg'
Learn More

L-Bornyl acetate, 95%

Product Code. 10217313 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10217313 2.5 kg 2.5kg
10254080 5 g 5g
10060570 100 g 100g
10599800 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10217313 Supplier Thermo Scientific Acros Supplier No. 106550025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5655-61-8 | C12H20O2 | 196.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5655-61-8
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00867808,MFCD00135942,MFCD00135942
InChI Key KGEKLUUHTZCSIP-HOSYDEDBSA-N
Synonym bornyl acetate, -bornyl acetate
PubChem CID 44630108
IUPAC Name [(1R,3S)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] acetate
SMILES CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C

Specifications

Color Colorless
Density 0.9875g/mL
Boiling Point 223°C to 224°C
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (as C12H20O2) Total esters (GC)
Refractive Index 1.4620 to 1.465
Quantity 2.5 kg
Beilstein 06,82
Merck Index 15,1341
Specific Gravity 0.9875
Specific Rotation Condition − 38.00 (20.00°C c=neat)
Specific Rotation -38
Solubility Information Solubility in water: .. Other solubilities: miscible with 95% alcohol and ether
Formula Weight 196.29
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material L-Bornyl acetate, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.