missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Borneol, 98.0%, MP Biomedicals™

Product Code. 11407781 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11407781 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11407781 Supplier MP Biomedicals Supplier No. 0215051380
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 464-45-9
Color White
Assay Percent Range 0.98
Molecular Formula C10H18O
MDL Number MFCD00003759,MFCD00066426,MFCD00066427
Quantity 100 g
Synonym --borneol, borneol, l-borneol, 1s,2r,4s-1,7,7-trimethylbicyclo 2.2.1 heptan-2-ol, linderol, 1s-endo---borneol, borneo camphor, bornyl alcohol, sumatra camphor, dl-borneol
InChI Key DTGKSKDOIYIVQL-NQMVMOMDSA-N
SMILES CC1(C)[C@@H]2CC[C@]1(C)[C@H](O)C2
IUPAC Name (1S,2R,4R)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-ol
Molecular Weight (g/mol) 154.25
PubChem CID 1201518
ChEBI CHEBI:15394
Formula Weight 154.3
Physical Form Crystalline Powder
Chemical Name or Material L-(-)-Borneol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.