missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Borneol, 97%

Product Code. 10215640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10215640 100 g Plastic bottle 100g
10023383 25 g Plastic bottle 25g
10184410 500 g Plastic bottle 500g
15189203 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10215640 Supplier Thermo Scientific Acros Supplier No. 160651000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 464-45-9 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 464-45-9
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00003759,MFCD00066426,MFCD00066427
InChI Key DTGKSKDOIYIVQL-NQMVMOMDSA-N
Synonym --borneol, borneol, l-borneol, 1s,2r,4s-1,7,7-trimethylbicyclo 2.2.1 heptan-2-ol, linderol, 1s-endo---borneol, borneo camphor, bornyl alcohol, sumatra camphor, dl-borneol
PubChem CID 1201518
ChEBI CHEBI:15394
IUPAC Name (1S,3R,4S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol
SMILES CC1(C)[C@@H]2CC[C@]1(C)[C@H](O)C2

Specifications

Melting Point 204°C to 208°C
Color White to Yellow
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 06,72
Merck Index 15,134
Specific Rotation -36.2
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 154.25
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(-)-Borneol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • L(-)-Borneol
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.