missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Aspartyl-L-phenylalanine, 95%

Product Code. 10073863 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass Bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10073863 500 mg Glass Bottle 500mg
10647061 100 mg Glass Bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10073863 Supplier Thermo Scientific Chemicals Supplier No. 310025000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13433-09-5 | C13H16N2O5 | 280.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13433-09-5
Molecular Formula C13H16N2O5
Molecular Weight (g/mol) 280.28
MDL Number MFCD00063155
InChI Key YZQCXOFQZKCETR-UWVGGRQHSA-N
Synonym l-aspartyl-l-phenylalanine, aspartyl-phenylalanine, demethylaspartame, aspartylphenylalanine, asp-phe, h-asp-phe-oh, l-a-aspartyl-l-phenylalanine, l-alpha-aspartyl-l-phenylalanine, alpha-aspartylphenylalanine, s-3-amino-4-s-1-carboxy-2-phenylethyl amino-4-oxobutanoic acid
PubChem CID 93078
ChEBI CHEBI:73830
SMILES N[C@@H](CC(O)=O)C(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O

Specifications

Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Linear Formula HO2CCH2CH(NH2)CONHCH(CH2C6H5)CO2H
Specific Rotation +10.0° to +13.0° (24°C, 589nm) (c=1, 0.5N HCl)
Water 3% max. (K.F.)
IUPAC Name (3S)-3-amino-4-[[(1S)-1-carboxy-2-phenylethyl]amino]-4-oxobutanoic acid
Specific Rotation Condition + 11.50 (24.00°C c=1,0.5 M HCl)
Quantity 500 mg
Formula Weight 280.28
Percent Purity ≥94% (HPLC)
Physical Form Powder
Chemical Name or Material L-Aspartyl-L-phenylalanine, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.