missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic Acid (White Crystals or Crystalline Powder), Fisher BioReagents

Product Code. 10796254
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10796254 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10796254 Supplier Fisher BioReagents Supplier No. BP374100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
This amino acid is suitable for use in tissue culture systems requiring additives.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 270°C
Ammonium 0.02% max.
CAS 56-84-8
pH 2.5 to 3.5
Color White
Heavy Metals (as Pb) 10ppm max.
Ignition Residue 0.1% max.
Identification Pass Test
Assay Percent Range 98.5 to 101.0 %
Loss on Drying 0.2% max.
Other Amino Acids Chromatographically not detectable
Molecular Formula C4H7NO4
Packaging Glass Bottle
MDL Number MFCD00002616
Specific Rotation 24.8 to 25.8° (+ or -)
Merck Index 15, 830
State of Solution (%T) 98.0% min.
Synonym l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid
Alpha Vector Aspartic Acid
InChI Key CKLJMWTZIZZHCS-UHFFFAOYNA-N
Density 1.6613g/cm³
SMILES NC(CC(O)=O)C(O)=O
IUPAC Name (2S)-2-aminobutanedioic acid
Molecular Weight (g/mol) 133.10
Quantity 100 g
PubChem CID 5960
ChEBI CHEBI:17053
Formula Weight 133.05
Percent Purity 98.5 to 101.0%
Physical Form Solid
Chemical Name or Material L-Aspartic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.