missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic Acid Sodium Salt Monohydrate, TRC

Product Code. 30179365
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179365 50 g 50g
30179026 5 g 5g
30179256 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179365 Supplier Toronto Research Chemicals Supplier No. TRCA78899350G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 323194-76-9
Molecular Formula C4H6NO4Na . H2O
Molecular Weight (g/mol) 173.1
Synonym L-Aspartic acid, sodium salt, hydrate (1:1:1), L-Aspartic acid, monosodium salt, monohydrate (9CI), Sodium L-aspartate monohydrate, Aspartic acid monosodium salt monohydrate, Monosodium L-aspartate monohydrate, Monosodium aspartate monohydrate, Sodium hydrogen (2S)-2-aminobutanedioate monohydrate
IUPAC Name sodium;(2S)-2-amino-4-hydroxy-4-oxobutanoate;hydrate
SMILES O.[Na+].N[C@@H](CC(=O)O)C(=O)[O-]

Specifications

InChI Formula InChI=1S/C4H7NO4.Na.H2O/c5-2(4(8)9)1-3(6)7;;/h2H,1,5H2,(H,6,7)(H,8,9);;1H2/q;+1;/p-1/t2-;;/m0../s1
Quantity 50 g
Formula Weight 173.03
Shipping Condition Room Temperature
Chemical Name or Material L-Aspartic Acid Sodium Salt Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.