missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic Acid, Pure, Fisher Chemical

Product Code. 10345550 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10345550 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10345550 Supplier Fisher Chemical Supplier No. A/8961/53

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 230°C
CAS 56-84-8
Color White
Molecular Formula C4H7NO4
Packaging Plastic Powder Jar
MDL Number 2616
InChI Key CKLJMWTZIZZHCS-UHFFFAOYNA-N
SMILES NC(CC(O)=O)C(O)=O
IUPAC Name 2-aminobutanedioic acid
Molecular Weight (g/mol) 133.10
Quantity 500 g
Formula Weight 133.1g/mol
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.