missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic acid, MedChemExpress

Product Code. 16492539
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492539 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16492539 Supplier Medchem Express Supplier No. HYN0666
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-Aspartic acid is is an amino acid, shown to be a suitable prodrug for colon-specific drug deliverly.

L-Aspartic acid is is an amino acid, shown to be a suitable prodrug for colon-specific drug deliverly.

Specifications

Chemical Name or Material L-Aspartic acid
CAS 56-84-8
Color White
Purity Grade Notes Research
Molecular Formula C4H7NO4
Solubility Information H2O : 8.33 mg/mL (62.58 mM; Need ultrasonic)
SMILES N[C@H](C(O)=O)CC(O)=O
Molecular Weight (g/mol) 133.1
Quantity 1 g
Formula Weight 133.1
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.