missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic acid di-tert-butyl ester hydrochloride, 98%

Product Code. p-7055196
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15418476 1 g 1g
15428476 5 g 5g
15438476 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15418476 Supplier Thermo Scientific Alfa Aesar Supplier No. H63716.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1791-13-5 | C12H24ClNO4 | 281.777 g/mol

L-Aspartic acid di-tert-butyl ester hydrochloride may be used in neurological comparison studies with structurally similar compounds such as L-Glutamic acid di-tert-butyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Aspartic acid di-tert-butyl ester hydrochloride may be used in neurological comparison studies with structurally similar compounds such as L-Glutamic acid di-tert-butyl ester.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1791-13-5
Molecular Formula C12H24ClNO4
Molecular Weight (g/mol) 281.777
MDL Number MFCD00034851
InChI Key GVLZIMQSYQDAHB-QRPNPIFTSA-N
Synonym h-asp otbu-otbu.hcl, l-aspartic acid di-tert-butyl ester hydrochloride, s-di-tert-butyl 2-aminosuccinate hydrochloride, h-asp otbu-otbu hcl, di-tert-butyl l-aspartate hydrochloride, l-aspartic acid, bis 1,1-dimethylethyl ester, hydrochloride, l-aspartic acid di-t-butyl ester hydrochloride, h-asp obut-obut·hcl, 1,4-di-tert-butyl 2s-2-aminobutanedioate hydrochloride, h-asp otbu-otbucl
PubChem CID 13768146
IUPAC Name ditert-butyl (2S)-2-aminobutanedioate;hydrochloride
SMILES CC(C)(C)OC(=O)CC(C(=O)OC(C)(C)C)N.Cl

Specifications

Beilstein 5652807
Quantity 1 g
Formula Weight 281.78
Percent Purity 98%
Physical Form Solid
Chemical Name or Material L-Aspartic acid di-tert-butyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.