Learn More
L-Aspartic acid, 99%, low metals content, Thermo Scientific Chemicals
£22.90 - £75.60
Chemical Identifiers
CAS | 56-84-8 |
---|---|
Molecular Formula | C4H7NO4 |
Molecular Weight (g/mol) | 133.10 |
MDL Number | MFCD00002616 |
InChI Key | CKLJMWTZIZZHCS-UHFFFAOYNA-N |
Synonym | l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid |
PubChem CID | 5960 |
ChEBI | CHEBI:17053 |
IUPAC Name | (2S)-2-aminobutanedioic acid |
SMILES | NC(CC(O)=O)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11319478
|
Thermo Scientific Alfa Aesar
043317.22 |
100 g |
£22.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11309478
|
Thermo Scientific Alfa Aesar
043317.36 |
500 g |
£75.60
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
L-Aspartic acid is used as a component of parenteral and enteral nutrition and as a pharmaceutical ingredient. it is used for cell culture and in manufacturing processes. It is widely utilized for mineral supplementation in the salt form.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
56-84-8 | |
133.10 | |
CKLJMWTZIZZHCS-UHFFFAOYNA-N | |
5960 | |
(2S)-2-aminobutanedioic acid |
C4H7NO4 | |
MFCD00002616 | |
l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid | |
CHEBI:17053 | |
NC(CC(O)=O)C(O)=O |
Specifications
>300°C (decomposition) | |
C4H7NO4 | |
1723530 | |
l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid | |
CKLJMWTZIZZHCS-UHFFFAOYNA-N | |
(2S)-2-aminobutanedioic acid | |
5960 | |
133.1 | |
1.66 g/mL | |
Powder |
56-84-8 | |
MFCD00002616 | |
14,840 | |
Soluble in water(4.5g/L). | |
NC(CC(O)=O)C(O)=O | |
133.10 | |
CHEBI:17053 | |
99% | |
Odorless | |
L-Aspartic acid, Low metals content |
Safety and Handling
missing translation for 'einecsNumber' : 200-291-6
missing translation for 'rtecsNumber' : CI9098500
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only