missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic Acid 4-tert-Butyl Ester, TRC

Product Code. 30179002
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179002 5 g 5g
30178940 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179002 Supplier Toronto Research Chemicals Supplier No. TRCA7889305G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3057-74-7
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
Synonym L-Aspartic Acid 4-(1,1-Dimethylethyl) Ester, (S)-2-Amino-4-tert-butoxy-4-oxobutanoic Acid, Aspartic Acid beta-tert-Butyl Ester, L-Aspartic Acid beta-tert-Butyl Ester, beta-tert-Butyl L-Aspartate, beta-tert-Butyl Aspartate
IUPAC Name (2S)-2-amino-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)CC(N)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H15NO4/c1-8(2,3)13-6(10)4-5(9)7(11)12/h5H,4,9H2,1-3H3,(H,11,12)/t5-/m0/s1
Formula Weight 189.1001
Chemical Name or Material L-Aspartic Acid 4-tert-Butyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.