missing translation for 'onlineSavingsMsg'
Learn More

L-Asparagine, TRC

Product Code. 30138063
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138063 5 g 5g
30137877 25 g 25g
30011398 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138063 Supplier Toronto Research Chemicals Supplier No. TRCA7900285G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-47-3
Molecular Formula C4 H8 N2 O3
Molecular Weight (g/mol) 132.12
Synonym L-Asparagine, Asparagine, L- (8CI), (-)-Asparagine, (S)-2,4-Diamino-4-oxobutanoic acid, (S)-Asparagine, Agedoite, Altheine, Asn, Asparagine, Asparagine acid, Asparamide, Aspartamic acid, Aspartic acid amide, Aspartic acid β-amide, Butanoic acid, 2,4-diamino-4-oxo-, (S)-, Crystal VI, L-2,4-Diamino-4-oxobutanoic acid, L-Aspartamine, L-β-Asparagine, NSC 82391, l-Asparagine, α-Aminosuccinamic acid
IUPAC Name (2S)-2,4-diamino-4-oxobutanoic acid
SMILES N[C@@H](CC(=O)N)C(=O)O

Specifications

InChI Formula InChI=1S/C4H8N2O3/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H2,6,7)(H,8,9)/t2-/m0/s1
Quantity 5 g
Formula Weight 132.0535
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-Asparagine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.