missing translation for 'onlineSavingsMsg'
Learn More

L-Asparagine Monohydrate, TRC

Product Code. 17959874
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17959874 1 g 1g
17909294 10 g 10g
17949114 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17959874 Supplier Toronto Research Chemicals Supplier No. TRCA7900051G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5794-13-8
Molecular Formula C4 H8 N2 O3 . H2 O
Molecular Weight (g/mol) 150.13
Synonym L-Asparagine, hydrate (1:1), Asparagine, monohydrate, L- (8CI), L-Asparagine, monohydrate (9CI), Asparagine monohydrate, (2S)-2,4-Diamino-4-oxobutanoic acid monohydrate, L-Asparagine monohydrate
IUPAC Name (2S)-2,4-diamino-4-oxobutanoic acid;hydrate
SMILES O.N[C@@H](CC(=O)N)C(=O)O

Specifications

InChI Formula InChI=1S/C4H8N2O3.H2O/c5-2(4(8)9)1-3(6)7;/h2H,1,5H2,(H2,6,7)(H,8,9);1H2/t2-;/m0./s1
Quantity 1 g
Formula Weight 150.0641
Shipping Condition Room Temperature
Chemical Name or Material Asparagine monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.