missing translation for 'onlineSavingsMsg'
Learn More

L-Asparagine Anhydrous MP Biomedicals

Product Code. 11393969 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393969 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11393969 Supplier MP Biomedicals Supplier No. 0210079480
 Restricted Item
Estimated Shipment: 12-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Anhydrous chemical for biochemical and laboratory use

Specifications

CAS 70-47-3
Molecular Formula C4H8N2O3
Synonym l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid
InChI Key DCXYFEDJOCDNAF-REOHCLBHSA-N
SMILES C(C(C(=O)O)N)C(=O)N
IUPAC Name (2S)-2,4-diamino-4-oxobutanoic acid
Molecular Weight (g/mol) 132.119
Quantity 100 g
PubChem CID 6267
ChEBI CHEBI:17196
Formula Weight 132.1
Grade Biochemical
Chemical Name or Material L-Asparagine, Anhydrous
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.