missing translation for 'onlineSavingsMsg'
Learn More

L-Asparagine, Anhydrous ≥98.5%, Ultrapure

Product Code. 15885088 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic Bottle
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15885088 100 g Plastic Bottle 100g
15865258 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15885088 Supplier Thermo Scientific Supplier No. J11590.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 70-47-3 | C4H8N2O3 | 132.119 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-47-3
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.119
MDL Number MFCD00064401
InChI Key DCXYFEDJOCDNAF-REOHCLBHSA-N
Synonym l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid
PubChem CID 6267
ChEBI CHEBI:17196
IUPAC Name (2 S)-2,4-diamino-4-oxobutanoic acid
SMILES C(C(C(=O)O)N)C(=O)N

Specifications

Melting Point 234°C to 236°C (decomposition)
Packaging Plastic Bottle
Linear Formula NH2COCH2CH(NH2)COOH
Beilstein 1723527
Quantity 100 g
Formula Weight 132.12
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material L-Asparagine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.