missing translation for 'onlineSavingsMsg'
Learn More

Melford L-Asparagine Anhydrous

Product Code. p-200087089
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
This item is not returnable. View return policy

Product Code. 30018043

Brand: Melford A500351000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-Asparagine Anhydrous is commonly used as a component in cell culture media.

L-Asparagine Anhydrous is commonly used as a component in cell culture media. It provides a vital nitrogen source for cell growth and is particularly important in the cultivation of certain cell types, especially in the production of recombinant proteins and antibodies.

L-Asparagine Anhydrous is utilized in laboratory research, particularly in studies related to cell biology, molecular biology, and biochemistry. Its role as an amino acid makes it valuable for understanding cellular processes.

Specifications

Chemical Name or Material L-Asparagine, Anhydrous
CAS 70-47-3
Molecular Formula C4H8N2O3
Quantity 1 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.