missing translation for 'onlineSavingsMsg'
Learn More

Melford L-Asparagine Anhydrous

Product Code. 30018043
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018043 1 kg 1kg
30018042 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018043 Supplier Melford Supplier No. A500351000.0
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

L-Asparagine Anhydrous is commonly used as a component in cell culture media.

L-Asparagine Anhydrous is commonly used as a component in cell culture media. It provides a vital nitrogen source for cell growth and is particularly important in the cultivation of certain cell types, especially in the production of recombinant proteins and antibodies.

L-Asparagine Anhydrous is utilized in laboratory research, particularly in studies related to cell biology, molecular biology, and biochemistry. Its role as an amino acid makes it valuable for understanding cellular processes.

Specifications

Chemical Name or Material L-Asparagine, Anhydrous
CAS 70-47-3
Molecular Formula C4H8N2O3
Quantity 1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.