missing translation for 'onlineSavingsMsg'
Learn More

L-Asparagine, 99% [Anhydrous], MP Biomedicals

Product Code. 11425082 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. Quantity unitSize
11425082 25 g 25g
11435082 100 g 100g
11445082 500 g 500g
3 options
This item is not returnable. View return policy

Product Code. 11425082

Brand: MP Biomedicals 0219463025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 70-47-3
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.119
InChI Key DCXYFEDJOCDNAF-REOHCLBHSA-N
Synonym l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid
PubChem CID 6267
ChEBI CHEBI:17196
IUPAC Name (2S)-2,4-diamino-4-oxobutanoic acid
SMILES C(C(C(=O)O)N)C(=O)N

Specifications

Quantity 25 g
Formula Weight 132.1
Grade Cell Culture
Physical Form Crystals
Chemical Name or Material L-Asparagine, 99% [Anhydrous]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.