missing translation for 'onlineSavingsMsg'
Learn More

L-Asparaginase Antibody, FITC, Novus Biologicals™

Product Code. 18416970 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416970 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416970 Supplier Novus Biologicals Supplier No. NBP178086F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

L-Asparaginase Polyclonal antibody specifically detects L-Asparaginase in E. coli samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen L-Asparaginase
Applications ELISA
Classification Polyclonal
Conjugate FITC
Dilution ELISA
Formulation PBS
Gene Alias AnsA, AnsB, Asparaginase, Colaspase, Cytoplasmic asparaginase I, L ASNase I, L ASNase II, L asparaginase 1, L asparaginase 2, L asparaginase I, L asparaginase II, L asparaginase II precursor, L asparagine amidohydrolase I, L asparagine amidohydrolase II
Host Species Rabbit
Immunogen L-L-Asparaginase [E. coli] (Uniprot: P00805)
Purification Method Multi-step
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species E. coli
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.