missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Asparaginase Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78086AF532
This item is not returnable.
View return policy
Description
L-Asparaginase Polyclonal antibody specifically detects L-Asparaginase in E. coli samples. It is validated for Western Blot, ELISA
Specifications
| L-Asparaginase | |
| Polyclonal | |
| Western Blot, ELISA | |
| AnsA, AnsB, Asparaginase, Colaspase, Cytoplasmic asparaginase I, L ASNase I, L ASNase II, L asparaginase 1, L asparaginase 2, L asparaginase I, L asparaginase II, L asparaginase II precursor, L asparagine amidohydrolase I, L asparagine amidohydrolase II | |
| L-L-Asparaginase [E. coli] (Uniprot: P00805) | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. |
| Western Blot, ELISA | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Multi-step | |
| RUO | |
| E. coli | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction