missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic Acid, TraceSELECT™, ≥99.0% (RT), Solstice

Product Code. 15680800
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15680800 100 g Poly bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15680800 Supplier Honeywell-Fluka Supplier No. 05878100G
 Restricted Item
Estimated Shipment: 21-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TraceSELECT™,≥99.9998% (metals basis)

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions
TRUSTED_SUSTAINABILITY

Specifications

CAS 50-81-7
Melting Point 190°C to 194°C
pH 1.0 to 2.0 (176g/L at 25°C)
Packaging Poly bottle
Molecular Formula C6H8O6
MDL Number MFCD00064328
Quantity 100 g
UN Number NONH for all modes of transport
Beilstein 84272
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
Solubility Information 176g/L at 20°C
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
Molecular Weight (g/mol) 176.12
PubChem CID 54670067
ChEBI CHEBI:29073
Formula Weight 176.12g/mol
Percent Purity ≥99.0% (RT)
Grade TraceSELECT
Chemical Name or Material L-Ascorbic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.