missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Ascorbic Acid Sodium Salt, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
500 g
1 kg
Unit Size:
1kg
25g
500g
Specifications
Specifications
| Color | White |
| Assay Percent Range | ≥99% |
| Quantity | 25 g |
| Formula Weight | 198.11 |
| Physical Form | Powder |
| Chemical Name or Material | L-Ascorbic Acid Sodium Salt, Antimicrobial, Antioxidant, Reductant for the isolation of chloroplasts |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction