missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic acid sodium salt, MedChemExpress

Product Code. 16482069
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482069 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16482069 Supplier Medchem Express Supplier No. HYB0166A
 Restricted Item
Estimated Shipment: 24-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-Ascorbic acid sodium salt (Sodium L-ascorbate), an electron donor, is an endogenous antioxidant agent. L-Ascorbic acid sodium salt inhibits selectively Cav3.2 channels with an IC50 of 6.5 μM. L-Ascorbic acid sodium salt is also a collagen deposition enhancer and an elastogenesis inhibitor.

L-Ascorbic acid sodium salt (Sodium L-ascorbate), an electron donor, is an endogenous antioxidant agent. L-Ascorbic acid sodium salt inhibits selectively Cav3.2 channels with an IC50 of 6.5 μM. L-Ascorbic acid sodium salt is also a collagen deposition enhancer and an elastogenesis inhibitor.

Specifications

Chemical Name or Material L-Ascorbic acid sodium salt
CAS 134-03-2
Color White
Purity Grade Notes Research
Molecular Formula C6H7NaO6
Quantity 1 g
Synonym Sodium L-ascorbateVitamin C sodium salt
Solubility Information H2O : 100 mg/mL (504.77 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (5.05 mM; Need ultrasonic)
SMILES O[C@@H](CO)[C@]1([H])C(O[Na])=C(O)C(O1)=O
Molecular Weight (g/mol) 198.11
Formula Weight 198.11
Percent Purity 97.0%
Grade Research
Physical Form Crystals
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.