missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic Acid, Mikromol™

Product Code. 17978014
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17978014 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17978014 Supplier Mikromol Supplier No. MM0166.00
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

L-Ascorbic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Ascorbic Acid
CAS 50-81-7
Analyte or Component Names L-Ascorbic Acid
Shipping Condition Room Temperature
Molecular Formula C6 H8 O6
Certifications/Compliance ISO 17034
Synonym L-Ascorbic Acid, (+)-Ascorbic acid, 100M, 3-Oxo-L-gulofuranolactone, 3-keto-L-Gulofuranolactone, Adenex, Allercorb, Antiscorbic vitamin, Antiscorbutic vitamin, Ascoltin, Ascorbajen, Ascorbic acid, Ascorbicap, Ascorbutina, Ascorell, Ascorin, Ascorteal, Ascorvit, C-L 6/PW, C-Quin, C-Vimin, Cantan, Cantaxin, Catavin C, Ce-Mi-Lin, Ce-Vi-Sol, Cebicure, Cebion, Cebione, Cecon, Cegiolan, Ceglion, Ceklin, Celaskon, Celin, Cell C, Cemagyl, Cenetone, Cereon, Cergona, Cescorbat, Cetamid, Cetane, Cetane-Caps TC, Cetebe, Cetemican, Cevalin, Cevarol, Cevatine, Cevex, Cevimin, Cevital, Cevitamic acid, Cevitamin, Cevitan, Cevitex, Cewin, Chewcee, Ciamin, Cinal, Cipca, Citrovit, Colascor, Concemin, Davitamon C, E 300, Hicee, Hybrin, IDO-C, Juvamine, Kangbingfeng, L-(+)-Ascorbic acid, L-3-Keto-threo-hexuronic acid lactone, L-Lyxoascorbic acid, L-Xyloascorbic acid, L-threo-Ascorbic acid, Laroscorbine, Lemascorb, Liqui-Cee, NSC 218455, NSC 33832, Neo-Valdrin, P 1110, Planavit C, Proscorbin, Redoxon, Ribena, Ronotec 100, Rontex 100, Rovimix C, Rovimix C-EC, Scorbu C, Secorbate, Suncoat VC 40, Testascorbic, VC 40, VC 97, Vasc, Vicelat, Vicin, Viforcit, Viscorin, Viscorin 100M, Vitace, Vitacee, Vitacimin, Vitacin, Vitamin C, Vitamisin, Vitascorbol, Vitashure 160, Xitix, L-threo-Hex-2-enonic acid, gamma-lactone
InChI Formula InChI=1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1
SMILES OC[C@H](O)[C@H]1OC(=O)C(=C1O)O
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
Molecular Weight (g/mol) 176.12
Quantity 500 mg
Formula Weight 176.0321 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.