missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Ascorbic Acid ACS AR, Macron Fine Chemicals™
macron
£640.00
Chemical Identifiers
CAS | 50-81-7 |
---|---|
Molecular Formula | C6H8O6 |
Molecular Weight (g/mol) | 176.12 |
MDL Number | MFCD00064328 |
InChI Key | CIWBSHSKHKDKBQ-DOMZIZNONA-N |
Synonym | l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin |
PubChem CID | 54670067 |
ChEBI | CHEBI:29073 |
SMILES | OC[C@H](O)[C@H]1OC(=O)C(O)=C1O |
Chemical Identifiers
50-81-7 | |
176.12 | |
CIWBSHSKHKDKBQ-DOMZIZNONA-N | |
54670067 | |
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O |
C6H8O6 | |
MFCD00064328 | |
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin | |
CHEBI:29073 |
Specifications
50-81-7 | |
MFCD00064328 | |
CIWBSHSKHKDKBQ-DOMZIZNONA-N | |
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one | |
54670067 | |
176.13 | |
L-Ascorbic Acid |
C6H8O6 | |
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin | |
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O | |
176.12 | |
CHEBI:29073 | |
ACS |