missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Ascorbic acid, ACS, 99+%, Thermo Scientific Chemicals

Product Code. 11393288
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is all important nutrient and antioxidant. L-ascorbic acid is frequently practiced as a processing aid or as a preservative in certain nutrients or food components. It is likewise utilized in the synthetic polymer industry, in photo processing, and in metal technology. Miscellaneous uses have been proposed in cosmetics, fibers, preservation of blood, preservation of cut plants, cleaning agents, and in assay reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
PubChem CID 54670067
ChEBI CHEBI:29073
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

Melting Point ∽191°C (decomposition)
Density 1.65 g/mL
Assay Percent Range >99+%
Quantity 25 g
Beilstein 84272
Solubility Information Soluble in water. Insoluble in ether,chloroform,benzene,petroleum ether,oils,fats and fat solvents. L-ascorbic acid means that the compound′s stereochemistry is related to that of the levorotatory enantiomer of glyceraldehyde.
Formula Weight 176.12
Grade ACS Reagent
Physical Form Powder
Chemical Name or Material L-Ascorbic acid, For analysis ACS

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'