missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Ascorbic acid, ACS, 99+%

Product Code. 11383288
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11383288

Brand: Thermo Scientific Alfa Aesar 036237.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50-81-7 | C6H8O6 | 176.12 g/mol

It is all important nutrient and antioxidant. L-ascorbic acid is frequently practiced as a processing aid or as a preservative in certain nutrients or food components. It is likewise utilized in the synthetic polymer industry, in photo processing, and in metal technology. Miscellaneous uses have been proposed in cosmetics, fibers, preservation of blood, preservation of cut plants, cleaning agents, and in assay reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
PubChem CID 54670067
ChEBI CHEBI:29073
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

Melting Point ∼191°C (decomposition)
Density 1.65
Quantity 100 g
Beilstein 84272
Merck Index 14,830
Solubility Information Soluble in water. Insoluble in ether,chloroform,benzene,petroleum ether,oils,fats and fat solvents. L-ascorbic acid means that the compound′s stereochemistry is related to that of the levorotatory enantiomer of glyceraldehyde.
Formula Weight 176.12
Percent Purity ≥99%
Grade ACS
Physical Form Powder
Chemical Name or Material L-(+)-Ascorbic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.