missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic Acid, 99+%, Extra Pure, SLR, Fisher Chemical™

Product Code. 11473523 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473523 500 g 500g
11458952 100 g 100g
11483523 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473523 Supplier Fisher Chemical Supplier No. A/8880/53
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

L-Ascorbic Acid, 99+%, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
MDL Number 64328

Specifications

Melting Point 190°C
Color White
pH 2.1 to 2.6
Packaging Plastic powder jar
Quantity 500 g
Formula Weight 176.13g/mol
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.