missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ L-Ascorbic acid

Description
TB4055-RMU is an Ancillary Material Grade version of product L-Ascorbic acid (Cat. No. 4055). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies. Find out more about our Ancillary Material Grade product range.
Specifications
Specifications
| Chemical Name or Material | 3-Oxo-L-gulofuranolactone |
| CAS | 50-81-7 |
| Quantity | 500 mg |
| Target | iPSC Generation |
| Molecular Formula | C6H8O6 |
| Purity | 99% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?